Please login to your online account to display your discounted pricing

Triiron dodecacarbonyl, 99%, stabilized, ACROS Organics™

$64.40 - $64.40

Chemical Identifiers

Chemical Name or Material Triiron dodecacarbonyl
Assay Percent Range 99%
Molecular Formula C12Fe3O12
Linear Formula Fe3(CO)12
Formula Weight 503.66
MDL Number MFCD00135617
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC209290050

  SDS
thermo fisher scientific
AC209290050
Glass bottle 5g Each for $64.40

AC209290250

  SDS
thermo fisher scientific
AC209290250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystals
Formula Weight 503.66
Quantity 5g
Color Black
Density 1.9900g/mL
Chemical Name or Material Triiron dodecacarbonyl
Packaging Glass bottle
Stabilizer 5 to 10% MeOH
Assay Percent Range 99%
Molecular Formula C12Fe3O12
Linear Formula Fe3(CO)12
MDL Number MFCD00135617
CAS 17685-52-8
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Flammable solid. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. May cause damage to organs.
Health Hazard 3 GHS P Statement Keep container tightly closed. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection.
Infrared Spectrum Authentic
Elemental Analysis 98.5% min.
Safety and Handling

Warning

GHS P Statement Keep container tightly closed. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection.

GHS P Statement Flammable solid. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. May cause damage to organs.