Trihalomethanes Mixture (For EPA 501)
$104.09 - $104.09
Chemical Identifiers
Chemical Name or Material Trihalomethanes Mixture
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
US-THM-501N-1 Ultra Scientific
THM501N1
Ampule 1 × 1mL Each for $104.09
Specifications Description & Specifications
Chemical Identifiers
Trihalomethanes Mixture
Specifications
1 × 1mL
For EPA Method 501
Trihalomethanes Mixture
Each component at 100μg/mL in Methanol:
Bromodichloromethane
Bromoform
Chloroform
Dibromochloromethane
Ampule