Please login to your online account to display your discounted pricing

Triformamide, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC380440250

 View more versions of this product

Catalog No. AC380440250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystals
Formula Weight 101.06
Quantity 25g
Color White
Melting Point 95.0° to 100.0°C
Chemical Name or Material Triformamide
Elemental Analysis 96.0% min. (ex%C)
Assay Percent Range 97%
Molecular Formula C3H3NO3
Linear Formula N(COH)3
MDL Number MFCD08059356
CAS 25891-31-0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Synonym N, N-Diformylformamide
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Triformamide
Assay Percent Range 97%
Molecular Formula C3H3NO3
Linear Formula N(COH)3
Formula Weight 101.06
MDL Number MFCD08059356
Synonym N, N-Diformylformamide
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents