missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triformamide, 97%, Thermo Scientific™

Chemical Identifiers

CAS 25891-31-0
Molecular Formula C3H3NO3
Molecular Weight (g/mol) 101.06
MDL Number MFCD08059356
InChI Key PFBAGGWJGZAGCG-UHFFFAOYSA-N
Synonym triformamide
PubChem CID 6419719
IUPAC Name N,N-diformylformamide
SMILES C(=O)N(C=O)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC380440010
SDS View Documents Promotion Details
Thermo Fisher Scientific
AC380440010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC380440050
SDS View Documents Promotion Details
Thermo Fisher Scientific
AC380440050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

25891-31-0
101.06
PFBAGGWJGZAGCG-UHFFFAOYSA-N
6419719
C(=O)N(C=O)C=O
C3H3NO3
MFCD08059356
triformamide
N,N-diformylformamide

Specifications

25891-31-0
100.0°C to 101.0°C
97%
C3H3NO3
MFCD08059356
triformamide
PFBAGGWJGZAGCG-UHFFFAOYSA-N
N,N-diformylformamide
6419719
Crystals
Triformamide
White
96.0% min. (ex%C)
Authentic
N(COH)3
Glass bottle
1g
C(=O)N(C=O)C=O
101.06
101.06
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Product Certifications

RUO – Research Use Only