missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triformamide, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC380440010

 View more versions of this product

Catalog No. AC380440010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Triformamide
Authentic
White
1g
97%
N(COH)3
triformamide
C(=O)N(C=O)C=O
101.06
101.06
97%
96.0% min. (ex%C)
Glass bottle
100.0°C to 101.0°C
25891-31-0
C3H3NO3
MFCD08059356
PFBAGGWJGZAGCG-UHFFFAOYSA-N
N,N-diformylformamide
6419719
Crystals

Chemical Identifiers

25891-31-0
101.06
PFBAGGWJGZAGCG-UHFFFAOYSA-N
6419719
C(=O)N(C=O)C=O
C3H3NO3
MFCD08059356
triformamide
N,N-diformylformamide
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents