missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Trifluoromethyl)trimethylsilane [Trifluoromethylating Reagent], TCI America™

Manufacturer:  TCI America T157025G

 View more versions of this product

Catalog No. T157025G



Specifications

Specifications

(Trifluoromethyl)trimethylsilane [Trifluoromethylating Reagent]
Yellow
81290-20-2
MFCD00145454
trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent
C[Si](C)(C)C(F)(F)F
142.196
142.20
≥97.0% (GC)
55°C
25g
C4H9F3Si
1993
MWKJTNBSKNUMFN-UHFFFAOYSA-N
trimethyl(trifluoromethyl)silane
552549
Clear Liquid at 20°C

Chemical Identifiers

81290-20-2
142.196
MWKJTNBSKNUMFN-UHFFFAOYSA-N
552549
C[Si](C)(C)C(F)(F)F
C4H9F3Si
MFCD00145454
trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent
trimethyl(trifluoromethyl)silane
Safety and Handling

Safety and Handling

TSCA : No

Documents