missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Trifluoromethyl)trimethylsilane, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC279720250

 View more versions of this product

Catalog No. AC279720250



Description

Description

Specifications

Specifications

(Trifluoromethyl)trimethylsilane
-17°C
Glass bottle
0.95
Colorless
96%
98.5
98.5% min. (GC)
(CH3)3SiCF3
15, 9851
MWKJTNBSKNUMFN-UHFFFAOYSA-N
trimethyl(trifluoromethyl)silane
552549
Liquid
0.9500g/mL
Authentic
1.3300 to 1.3320
54.0°C to 55.0°C
25mL
81290-20-2
100.0
C4H9F3Si
MFCD00145454
ruppert's reagent, ruppert-prakash reagent, silane, trimethyl trifluoromethyl, tfmtms, tms-cf3, trifluoromethyl trimethylsilane, trifluoromethyltrimethylsilane, trimethyl trifluoromethyl silane, trimethylsilyl trifluoromethane, unii-a009786qpj
C[Si](C)(C)C(F)(F)F
142.2
142.2
99%

Chemical Identifiers

81290-20-2
142.2
MWKJTNBSKNUMFN-UHFFFAOYSA-N
552549
C[Si](C)(C)C(F)(F)F
C4H9F3Si
MFCD00145454
ruppert's reagent, ruppert-prakash reagent, silane, trimethyl trifluoromethyl, tfmtms, tms-cf3, trifluoromethyl trimethylsilane, trifluoromethyltrimethylsilane, trimethyl trifluoromethyl silane, trimethylsilyl trifluoromethane, unii-a009786qpj
trimethyl(trifluoromethyl)silane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA

SDS Documents