missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trifluoromethanesulfonic anhydride, 98%

Manufacturer:  Alfa Aesar™ A1176722

 View more versions of this product

Catalog No. AAA1176722



Description

Description

Applications
Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.

Solubility
Miscible with dichloromethane. Immiscible with hydrocarbons.

Notes
Moisture sensitive. Incompatible with water, strong bases, strong oxidizing agents, acids and alcohols.
Specifications

Specifications

Trifluoromethanesulfonic anhydride
-80°C
358-23-6
(CF3SO2)2O
UN3265
trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride
C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F
282.127
CHEBI:48509
98%
81°C to 83°C
100g
C2F6O5S2
MFCD00000408
1813600
WJKHJLXJJJATHN-UHFFFAOYSA-N
trifluoromethylsulfonyl trifluoromethanesulfonate
67749
282.13

Chemical Identifiers

358-23-6
282.127
WJKHJLXJJJATHN-UHFFFAOYSA-N
67749
trifluoromethylsulfonyl trifluoromethanesulfonate
C2F6O5S2
MFCD00000408
trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride
CHEBI:48509
C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : PB2772000

TSCA : TSCA

SDS Documents