missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trifluoromethanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals

Trifluoromethanesulfonic acid, 99%, CHF3O3S, CAS Number-1493-13-6, hotf, methanesulfonic acid, trifluoro, tfmsa, perfluoromethanesulfonic acid, tfoh, trifluoromethanesulphonic acid, triflic acid, trifluoromethane sulfonic acid, trimsylate, trifluoromethylsulfonic acid, 250mL, 03,IV,34, 162.0 deg.C

Supplier:  Thermo Scientific Chemicals 169890010

 View more versions of this product

Catalog No. AC169890010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Trifluoromethanesulfonic acid
-40.0°C
1.6900g/mL
Authentic
Glass bottle
1.3260 to 1.3300
MFCD00007514
03,IV,34
1.69
triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
ITMCEJHCFYSIIV-UHFFFAOYSA-N
trifluoromethanesulfonic acid
62406
CHEBI:48511
99%
Fuming Liquid
1493-13-6
Colorless to Yellow
162.0°C
99%
CHF3O3S
CF3SO3H
1 mL
04,532; 05,701; 06,617; 08,504; 09,531; 14,152; 15,41; 16,357; 17,24
15,9842
Solubility in water: soluble. Other solubilities: soluble in many polar organic solvents such as dim,sulfolane,dimethyl sulfoxide,dimethyl sulfone an
OS(=O)(=O)C(F)(F)F
150.07
2.87 mPa.s (25°C)
150.07
Extra Pure

Chemical Identifiers

1493-13-6
150.07
ITMCEJHCFYSIIV-UHFFFAOYSA-N
62406
OS(=O)(=O)C(F)(F)F
CHF3O3S
MFCD00007514
triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
CHEBI:48511
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause respiratory irritation.
May be corrosive to metals.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 216-087-5

Recommended Storage : May slightly darken during storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only