missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trifluoromethanesulfonic acid, 98+%

Manufacturer:  Alfa Aesar™ A1017318*

 View more versions of this product

Catalog No. AAA1017318



Description

Description

Applications
Trifluoromethanesulfonic acid acts as a catalyst for esterification reactions and an acidic titrant in nonaqueous acid-base titration. It is useful in protonations due to the presence of conjugate base triflate is non nucleophilic. It serves as a deglycosylation agent for glycoproteins. In addition, it is a precursor and a catalyst in organic chemistry. It reacts with acyl halides to prepare mixed triflate anhydrides, which are strong acylating agents used in Friedel-Crafts reactions. It acts as a key starting material for the preparation of ethers and olefins by reacting with alcohols as well as to prepare trifluoromethanesulfonic anhydride by dehydration reaction.

Solubility
Miscible with water, dimethyl sulfoxide, dimethyl formamide and acetonitrile.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong bases, water and metals.
Specifications

Specifications

Trifluoromethanesulfonic acid
-40°C
1493-13-6
≥98%
CF3SO3H
UN3265
14,9676
ITMCEJHCFYSIIV-UHFFFAOYSA-N
trifluoromethanesulfonic acid
62406
150.07
161°C to 162°C
50g
Pungent
CHF3O3S
MFCD00007514
1812100
Triflic acid
C(F)(F)(F)S(=O)(=O)O
150.071
CHEBI:48511

Chemical Identifiers

1493-13-6
150.071
ITMCEJHCFYSIIV-UHFFFAOYSA-N
62406
trifluoromethanesulfonic acid
CHF3O3S
MFCD00007514
Triflic acid
CHEBI:48511
C(F)(F)(F)S(=O)(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : PB2771000

TSCA : TSCA

SDS Documents