Please login to your online account to display your discounted pricing

Alfa Aesar™ Trifluoroiodomethane, 99.9%

Chemical Identifiers

Chemical Name or Material Trifluoroiodomethane
CAS 2314-97-8
Formula Weight 195.91
MDL Number MFCD00001060
UN Number UN1956
Beilstein 1732740
Synonym Iodotrifluoromethane; Trifluoromethyl iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3966530

  SDS
alfa aesar™
3966530
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.91
Chemical Name or Material Trifluoroiodomethane
TSCA TSCA
Packaging Cylinder
Refractive Index 1.3790
Physical Form Gas
Assay 99.9%
MDL Number MFCD00001060
CAS 2314-97-8
UN Number UN1956
Beilstein 1732740
Synonym Iodotrifluoromethane; Trifluoromethyl iodide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H280-H341
Contains gas under pressure
may explode if heated.
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement
P281-P201-P308+P313-P405-P410+P403-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Protect from sunlight.
Store in a well-ventilated place.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 2; Proper Shipping Name: COMPRESSED GAS N.O.S.
RTECS Number PB6975000
Density 2.36g/mL
Sensitivity Light Sensitive
Boiling Point −22.5°C
Quantity 250g
Safety and Handling

Warning

GHS P Statement
P281-P201-P308+P313-P405-P410+P403-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Protect from sunlight.
Store in a well-ventilated place.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H280-H341
Contains gas under pressure
may explode if heated.
Suspected of causing genetic defects.