missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(Trifluoroacetyl)benzotriazole (mixture of 1H- and 2H- isomers) 98.0+%, TCI America™
Supplier: TCI America T31741G
Specifications
(Trifluoroacetyl)benzotriazole (mixture of 1H- and 2H- isomers) | |
69°C | |
C8H4F3N3O | |
1 g | |
C1=CC=C2C(=C1)N=NN2C(=O)C(F)(F)F | |
215.135 | |
215.14 | |
Crystalline Powder |
183266-61-7 | |
White | |
MFCD00593044 | |
GVQIQOIKWUOEJP-UHFFFAOYSA-N | |
1-(benzotriazol-1-yl)-2,2,2-trifluoroethanone | |
3358778 | |
≥98.0% (HPLC) |
Chemical Identifiers
183266-61-7 | |
215.135 | |
GVQIQOIKWUOEJP-UHFFFAOYSA-N | |
1-(benzotriazol-1-yl)-2,2,2-trifluoroethanone |
C8H4F3N3O | |
MFCD00593044 | |
3358778 | |
C1=CC=C2C(=C1)N=NN2C(=O)C(F)(F)F |
Safety and Handling
TSCA : No