Learn More
Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals
Trifluoroacetic anhydride, 99+%, Acros Organics, Quantity: 500g, Packaging: Glass Bottle, Boiling Point: 39.1 deg.C, Undesignated, Melting Point: -63.5 deg.C, Molecular Weight: 210.03, Percent Purity: 99+%, Assay Percent Range: 99% min. (GC), Beilstein: 02, II, 186, CAS: 407-25-0
Supplier: Thermo Scientific Chemicals 147811000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagentSpecifications
Trifluoroacetic anhydride | |
-63.5°C | |
39.1°C | |
99+% | |
C4F6O3 | |
(CF3CO)2O | |
100 g | |
01,1221; 03,308; 05,701; 06,616; 07,389; 09,484; 12,530; 14,151; 15,14; 01,373 | |
Solubility in water: hydrolysis. Other solubilities: miscible with polar organic solvents, partically soluble in unpolar hydrocarbons | |
FC(F)(F)C(=O)OC(=O)C(F)(F)F | |
210.03 | |
210.03 | |
≥99% |
407-25-0 | |
1.5100g/mL | |
Authentic | |
Glass Bottle | |
1.3 | |
MFCD00000416 | |
02, II, 186 | |
trifluoroacetic anhydride, trifluoroacetic acid anhydride, 2,2,2-trifluoroacetic anhydride, bis trifluoroacetic anhydride, acetic acid, trifluoro-, anhydride, perfluoroacetic anhydride, trifluoroacetyl anhydride, tfaa, hexafluoroacetic anhydride, anhydrid kyseliny trifluoroctove | |
QAEDZJGFFMLHHQ-UHFFFAOYSA-N | |
(2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate | |
9845 | |
141mbar at 20°C | |
Liquid |
Chemical Identifiers
407-25-0 | |
210.03 | |
QAEDZJGFFMLHHQ-UHFFFAOYSA-N | |
9845 | |
FC(F)(F)C(=O)OC(=O)C(F)(F)F |
C4F6O3 | |
MFCD00000416 | |
trifluoroacetic anhydride, trifluoroacetic acid anhydride, 2,2,2-trifluoroacetic anhydride, bis trifluoroacetic anhydride, acetic acid, trifluoro-, anhydride, perfluoroacetic anhydride, trifluoroacetyl anhydride, tfaa, hexafluoroacetic anhydride, anhydrid kyseliny trifluoroctove | |
(2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only