Please login to your online account to display your discounted pricing

Trifluoroacetic Acid, Silver Salt 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  139730050

 View more versions of this product

Catalog No. AC139730050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5% (NH4SCN-titration)
Formula Weight 220.89
Quantity 5g
Color Light beige to gray
TSCA TSCA
EINECS Number 221-004-0
Fieser 01,1018; 07,323; 08,444; 11,471; 16,301; 17,154
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C2AgF3O2
Chemical Name or Material Trifluoroacetic acid, silver salt
MDL Number MFCD00013199
CAS 2966-50-9
Linear Formula CF3CO2Ag
Assay Percent Range 98%
Name Note 98%
Synonym Silver trifluoroacetate
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Very toxic to aquatic life.

Specifications

Chemical Name or Material Trifluoroacetic acid, silver salt
Name Note 98%
CAS 2966-50-9
Assay ≥97.5% (NH4SCN-titration)
Assay Percent Range 98%
Molecular Formula C2AgF3O2
Linear Formula CF3CO2Ag
Formula Weight 220.89
MDL Number MFCD00013199
Synonym Silver trifluoroacetate
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Very toxic to aquatic life.

SDS & Safety Documents