Please login to your online account to display your discounted pricing

Trifluoroacetic Acid, OR, Macron Fine Chemicals™


macron

Quantity 100g

Manufacturer: avantor performance  194257

 View more versions of this product

Catalog No. MK1942100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 100g
Physical Form Fuming Liquid
Assay 99% min.
Formula Weight 114.03
Color Colorless to light straw
Chemical Name or Material Trifluoroacetic Acid
Molecular Formula F3CCOOH
CAS 76-05-1

Specifications

Chemical Name or Material Trifluoroacetic Acid
CAS 76-05-1
Physical Form Fuming Liquid
Molecular Formula F3CCOOH
Formula Weight 114.03