missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trifluoroacetic Acid, OR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 76-05-1
Physical Form Fuming Liquid
Molecular Formula F3CCOOH
Formula Weight 114.03
Chemical Name or Material Trifluoroacetic Acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MK1942100 Avantor Performance
194257
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK1942500 Avantor Performance
194259*
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

76-05-1
F3CCOOH
Trifluoroacetic Acid
Fuming Liquid
114.03

Specifications

100g
Fuming Liquid
F3CCOOH
Trifluoroacetic Acid
114.03
Colorless to light straw
76-05-1
99% min.