Please login to your online account to display your discounted pricing

Trifluoroacetic Acid, HPLC Grade, J.T.Baker™


High purity solvents for use in analytical and preparative separations

Quantity 10 x 1mL
Packaging Glass Ampoule

Manufacturer: avantor performance  947000

 View more versions of this product

Catalog No. 02004494

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 114.03
Name Note Gradient Tested for HPLC Peptide and Protein Analysis
Specific Gravity 1L = 1.53kg
Chemical Name or Material Trifluoroacetic Acid
CAS 76-05-1
Molecular Formula F3CCOOH
Packaging Glass Ampoule
Purity Grade HPLC Grade
Quantity 10 x 1mL

Specifications

Chemical Name or Material Trifluoroacetic Acid
CAS 76-05-1
Name Note Gradient Tested for HPLC Peptide and Protein Analysis
Purity Grade HPLC Grade
Molecular Formula F3CCOOH
Formula Weight 114.03