missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trifluoroacetic acid, HPLC Grade, >99.5%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 44630AY

 View more versions of this product

Catalog No. AA44630AY



Specifications

Specifications

Trifluoroacetic Acid
Pungent
Hygroscopic
72°C to 73°C
-15°C
Water 0.1%
>99.5%
MFCD00004169
742035
Miscible with water,ether,acetone,ethanol,benzene,hexane and carbon tetrachloride.
C(=O)(C(F)(F)F)O
114.023
CHEBI:45892
Liquid
1.48
1.284
8mm Hg at 20°C
Colorless
4 x 500mL
76-05-1
C2HF3O2
2699
acetic acid, trifluoro
DTQVDTLACAAQTR-UHFFFAOYSA-N
2,2,2-trifluoroacetic acid
6422
114.02
HPLC

Chemical Identifiers

76-05-1
114.023
DTQVDTLACAAQTR-UHFFFAOYSA-N
6422
2,2,2-trifluoroacetic acid
C2HF3O2
MFCD00004169
acetic acid, trifluoro
CHEBI:45892
C(=O)(C(F)(F)F)O
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H332-H335

DOTInformation : DOT Class: 8, Packaging Group: I

EINECSNumber : 200-929-3

RTECSNumber : AJ9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents