missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trifluoroacetic acid-d, for NMR, 99.5% atom D, ACROS Organics™

$112.56 - $223.12

Chemical Identifiers

CAS 599-00-8
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 115.029
MDL Number MFCD00044563
InChI Key DTQVDTLACAAQTR-DYCDLGHISA-N
Synonym acetic acid-d, 2,2,2-trifluoro, acetic acid-d, trifluoro, acetic acid-d,2,2,2-trifluoro, cf3cood, deuterotrifluoroacetic acid, tfa-d, trifluoroacetic 2h acid, trifluoroacetic acid-d, trifluoroacetic acid-d tfa-d, trifluoroacetic acid-d, 99.5 atom % d
PubChem CID 71502
IUPAC Name deuterio 2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC325310100
SDS View Documents
Thermo Fisher Scientific
325310100
10mL Glass bottle
Each for $112.56
 
AC325310250
SDS View Documents
Thermo Fisher Scientific
325310250
25mL Glass bottle
Each for $223.12
 
Description

Description

For NMR
Specifications

Chemical Identifiers

599-00-8
115.029
DTQVDTLACAAQTR-DYCDLGHISA-N
71502
C(=O)(C(F)(F)F)O
C2HF3O2
MFCD00044563
acetic acid-d, 2,2,2-trifluoro, acetic acid-d, trifluoro, acetic acid-d,2,2,2-trifluoro, cf3cood, deuterotrifluoroacetic acid, tfa-d, trifluoroacetic 2h acid, trifluoroacetic acid-d, trifluoroacetic acid-d tfa-d, trifluoroacetic acid-d, 99.5 atom % d
deuterio 2,2,2-trifluoroacetate

Specifications

1.5000g/mL
Glass bottle
Colorless
599-00-8
-15.0°C
CF3CO2D
acetic acid-d, 2,2,2-trifluoro, acetic acid-d, trifluoro, acetic acid-d,2,2,2-trifluoro, cf3cood, deuterotrifluoroacetic acid, tfa-d, trifluoroacetic 2h acid, trifluoroacetic acid-d, trifluoroacetic acid-d tfa-d, trifluoroacetic acid-d, 99.5 atom % d
C(=O)(C(F)(F)F)O
115.029
115.03
NMR
99.4% min.
0.05% max. (Coulometric)
10mL
75.0°C
C2HF3O2
MFCD00044563
DTQVDTLACAAQTR-DYCDLGHISA-N
deuterio 2,2,2-trifluoroacetate
71502
Liquid
Trifluoroacetic acid-d, 99.5 atom % D, for NMR
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS