Please login to your online account to display your discounted pricing

Alfa Aesar™ Trifluoroacetic acid-d, 99.5%(Isotopic)

Chemical Identifiers

Chemical Name or Material Trifluoroacetic acid-d
CAS 599-00-8
Name Note Isotopic
Formula Weight 115.03
MDL Number MFCD00044563
UN Number UN2699
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4235709

  SDS
alfa aesar™
4235709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4235714

  SDS
alfa aesar™
4235714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 115.03
Packaging Prescored Ampules
Density 1.50g/mL
Assay 99.5%
Odor Acrid
Physical Form Liquid
Health Hazard 1 Danger
Melting Point −15°C
Chemical Name or Material Trifluoroacetic acid-d
Name Note Isotopic
TSCA TSCA
Boiling Point 72°C
MDL Number MFCD00044563
Health Hazard 2 GHS H Statement
H314-H318-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
UN Number UN2699
DOT Information Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: TRIFLUOROACETIC ACID
CAS 599-00-8
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.