Please login to your online account to display your discounted pricing

Trifluoroacetic Acid (Clear, Fuming Liq./Biotechnology Grade), Fisher Chemical

$794.69 - $794.69

Chemical Identifiers

Chemical Name or Material Trifluoroacetic Acid
CAS 76-05-1
Purity Grade Biotech Grade
Physical Form Liquid
Assay ≥98.5%
Assay Percent Range ≥98.5 %
Molecular Formula C2HF3O2
Linear Formula CF3COOH
Formula Weight 114.02
MDL Number MFCD00004169
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

O6115-100

  SDS
fisher scientific company
O6115100
Glass Bottle 100mL Each for $794.69
Description & Specifications

Specifications

Assay ≥98.5%
Melting Point −15°C
Boiling Point 72°C
Purity Grade Biotech Grade
Formula Weight 114.02
Quantity 100mL
DOT Information DOT Class 8, : Corrosive
Chemical Name or Material Trifluoroacetic Acid
Sulfate 10ppm max.
Vapor Pressure 107mbar at 25°C
Water 0.05% max.
pH 0.45
Color Clear Colorless
Physical Form Liquid
Assay Percent Range ≥98.5 %
Chloride 500ppm max.
Molecular Formula C2HF3O2
Linear Formula CF3COOH
MDL Number MFCD00004169
CAS 76-05-1
ChemAlert Storage Symbol White
Density 1.535g/cm³
Fluoride 50ppm max.
Health Hazard 2 DANGER!
Infrared Spectrum Conforms to reference
Health Hazard 3 Emergency Overview
Causes severe burns by all exposure routes. Inhalation may cause central nervous system effects. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:0
Instability:0
Packaging Glass Bottle
Additional Information Vapor Pressure: 107mbar at 25°C

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Safety and Handling
 

Emergency Overview
Causes severe burns by all exposure routes. Inhalation may cause central nervous system effects. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:0
Instability:0

DANGER!