Learn More
Trifluoroacetic acid, 99+%, extra pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 139720100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Trifluoroacetic acid | |
1.5350g/mL | |
Conforms to Reference | |
Plastic drum | |
1.3 | |
MFCD00004169 | |
02, II, 186 | |
1.535 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoracetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
2,2,2-trifluoroacetic acid | |
6422 | |
CHEBI:45892 | |
107mbar at 25°C | |
Extra Pure |
76-05-1 | |
72.0°C | |
99% | |
C2HF3O2 | |
CF3CO2H | |
10kg | |
01,1219; 02,433; 03,305; 04,530; 05,695; 06,613; 07,388; 08,503; 09,483; 10,418; 11,557; 12,529; 14,322; 15,338; 16,83; 17,340 | |
15, 9847 | |
Solubility in water: miscible. Other solubilities: miscible with ether, acetone, ethanol, benzene,, ccl4, hexane, methanol, xylene and hexane | |
C(=O)(C(F)(F)F)O | |
114.02 | |
0.813 mPa.s (25°C) | |
114.02 | |
99% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 200-929-3
RUO â Research Use Only