missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trifluoroacetic Acid, 99%, Alfa Aesar™

Solvent, catalyst, strong non-oxidizing acid

Manufacturer:  Alfa Aesar A1219822*

 View more versions of this product

Catalog No. AAA1219822


 

Description

Description

Applications
The combination of properties like solubility in most of the solvents, volatility, catalytic property, and strong acidity with non-oxidizing nature makes it a widely used reagent in organic synthesis. Trifluoroacetic acid is an important building block in the synthesis of pharmaceuticals, agrochemicals and performance products. It is a precursor to many fluorinated compounds, and widely used in peptide synthesis and other organic transformations involving deprotection of t-BOC group. TFA finds use as an ion pairing agent in liquid chromatography, as a solvent in NMR spectroscopy and as a calibrant in mass spectrometry.

Solubility
Miscible with ether, acetone, ethanol, benzene, hexane, and CCl4

Notes
Trifluoroacetic acid (TFA) is a stable, hygroscopic, volatile and low-boiling organic acid. Storage in a cool place is recommended.
Specifications

Specifications

Trifluoroacetic Acid
99%
1.480
C(=O)(C(F)(F)F)O
114.023
CHEBI:45892
Liquid
72°C to 73°C
76-05-1
C2HF3O2
DTQVDTLACAAQTR-UHFFFAOYSA-N
2,2,2-trifluoroacetic acid
6422
114.02
-15°C
100g

Chemical Identifiers

76-05-1
114.023
6422
C2HF3O2
DTQVDTLACAAQTR-UHFFFAOYSA-N
CHEBI:45892
Safety and Handling

Safety and Handling

storageNote1 : Hygroscopic

SDS Documents