missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trifluoroacetic acid, 99.5+%, Alfa Aesar™

Manufacturer:  Alfa Aesar 31771A4

 View more versions of this product

Catalog No. AA31771A4



Specifications

Specifications

Trifluoroacetic acid
>99.5%
CF3CO2H
UN2699
14,9681
1.480g/mL
C(=O)(C(F)(F)F)O
114.023
CHEBI:45892
Liquid
Pungent
-15°C
72°C to 73°C
Hygroscopic
76-05-1
C2HF3O2
MFCD00004169
742035
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoracetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
DTQVDTLACAAQTR-UHFFFAOYSA-N
2,2,2-trifluoroacetic acid
6422
114.02
≥99.5%
1.2840
Colorless
2.5kg

Chemical Identifiers

76-05-1
114.023
DTQVDTLACAAQTR-UHFFFAOYSA-N
6422
2,2,2-trifluoroacetic acid
C2HF3O2
MFCD00004169
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoracetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
CHEBI:45892
C(=O)(C(F)(F)F)O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H332
Causes severe skin burns and eye damage.
Harmful if inhaled.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: TRIFLUOROACETIC ACID

RTECSNumber : AJ9625000

TSCA : TSCA

SDS Documents