missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Trifluoroacetamidine, tech. 85%

Manufacturer:  Alfa Aesar™ L1242222

 View more versions of this product

Catalog No. AAL1242222



Description

Description

Applications
Trifluoroacetamidine is used as probe for determination of membrane potential and extra/intracellular volume of erythrocytes by fluorine-19 NMR studies. It is a reagent used as protected ammonia equivalent.

Solubility
Fully miscible in water.

Notes
Store at -20°C. Incompatible with oxidizing agents. Moisture Sensitive.
Specifications

Specifications

Trifluoroacetamidine
100g
C2H3F3N2
UN3267
trifluoroacetamidine, trifluoroethanimidamide, 2,2,2-trifluoroacetamidine, ethanimidamide, 2,2,2-trifluoro, sntljqhjptadtaetp@, trifluoroacetoamidine, trifluoroacetamideine, trifluoro-acetamidine, acmc-20aje4, 2,2,2-trifluoroacetoamidine
C(=N)(C(F)(F)F)N
112.055
112.05
Technical
35°C to 37°C (11mmHg)
354-37-0
MFCD00041545
605978
NITMACBPVVUGOJ-UHFFFAOYSA-N
2,2,2-trifluoroethanimidamide
2776882
85%

Chemical Identifiers

354-37-0
112.055
NITMACBPVVUGOJ-UHFFFAOYSA-N
2776882
C(=N)(C(F)(F)F)N
C2H3F3N2
MFCD00041545
trifluoroacetamidine, trifluoroethanimidamide, 2,2,2-trifluoroacetamidine, ethanimidamide, 2,2,2-trifluoro, sntljqhjptadtaetp@, trifluoroacetoamidine, trifluoroacetamideine, trifluoro-acetamidine, acmc-20aje4, 2,2,2-trifluoroacetoamidine
2,2,2-trifluoroethanimidamide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

Recommended Storage : −30° to −10°C

Documents