Learn More
Trifluoroacetaldehyde methyl hemiacetal, 90%, technical, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC449131000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Trifluoroacetaldehyde methyl hemiacetal | |
1.3600g/mL | |
42°C | |
C3H5F3O2 | |
MFCD00013572 | |
1-methoxy-2,2,2-trifluoroethanol, 2,2,2-erifluoro-1-methoxyethanol, ethanol, 1-methoxy-2,2,2-trifluoro, ethanol, 2,2,2-trifluoro-1-methoxy, fluoral hemimethylacetal, fluoral methyl hemiacetal, trifluoroacetaldehyde hemi methyl acetal, trifluoroacetaldehyde methyl hemiacetal, trifluoroacetaldehyde methyl hemiacetal, tech., zlchem 1308 | |
GWTBCUWZAVMAQV-UHFFFAOYNA-N | |
COC(O)C(F)(F)F | |
130.07 | |
130.07 | |
Technical |
431-46-9 | |
104.0°C to 106.0°C | |
88% min. (GC) | |
CF3CH(OCH3)OH | |
Glass bottle | |
100g | |
1.36 | |
2,2,2-trifluoro-1-methoxyethanol | |
9892 | |
90% |
Chemical Identifiers
431-46-9 | |
130.07 | |
GWTBCUWZAVMAQV-UHFFFAOYNA-N | |
9892 | |
COC(O)C(F)(F)F |
C3H5F3O2 | |
MFCD00013572 | |
1-methoxy-2,2,2-trifluoroethanol, 2,2,2-erifluoro-1-methoxyethanol, ethanol, 1-methoxy-2,2,2-trifluoro, ethanol, 2,2,2-trifluoro-1-methoxy, fluoral hemimethylacetal, fluoral methyl hemiacetal, trifluoroacetaldehyde hemi methyl acetal, trifluoroacetaldehyde methyl hemiacetal, trifluoroacetaldehyde methyl hemiacetal, tech., zlchem 1308 | |
2,2,2-trifluoro-1-methoxyethanol |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Warning
EINECSNumber : 207-072-4
RUO – Research Use Only