missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trifluoroacetaldehyde methyl hemiacetal, 90%, technical, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC449130050
Specifications
Trifluoroacetaldehyde methyl hemiacetal | |
42°C | |
1.36 | |
5g | |
88% min. (GC) | |
CF3CH(OCH3)OH | |
GWTBCUWZAVMAQV-UHFFFAOYSA-N | |
2,2,2-trifluoro-1-methoxyethanol | |
9892 | |
90% |
1.3600g/mL | |
Glass bottle | |
104.0°C to 106.0°C | |
431-46-9 | |
C3H5F3O2 | |
1-methoxy-2,2,2-trifluoroethanol, 2,2,2-erifluoro-1-methoxyethanol, ethanol, 1-methoxy-2,2,2-trifluoro, ethanol, 2,2,2-trifluoro-1-methoxy, fluoral hemimethylacetal, fluoral methyl hemiacetal, trifluoroacetaldehyde hemi methyl acetal, trifluoroacetaldehyde methyl hemiacetal, trifluoroacetaldehyde methyl hemiacetal, tech., zlchem 1308 | |
COC(C(F)(F)F)O | |
130.07 | |
130.07 | |
Technical |
Chemical Identifiers
431-46-9 | |
130.07 | |
1-methoxy-2,2,2-trifluoroethanol, 2,2,2-erifluoro-1-methoxyethanol, ethanol, 1-methoxy-2,2,2-trifluoro, ethanol, 2,2,2-trifluoro-1-methoxy, fluoral hemimethylacetal, fluoral methyl hemiacetal, trifluoroacetaldehyde hemi methyl acetal, trifluoroacetaldehyde methyl hemiacetal, trifluoroacetaldehyde methyl hemiacetal, tech., zlchem 1308 | |
2,2,2-trifluoro-1-methoxyethanol |
C3H5F3O2 | |
GWTBCUWZAVMAQV-UHFFFAOYSA-N | |
9892 | |
COC(C(F)(F)F)O |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Warning
EINECSNumber : 207-072-4