Please login to your online account to display your discounted pricing

Alfa Aesar™ Trifluoroacetaldehyde hydrate, tech., ca 75% in water

$123.41 - $123.41

Chemical Identifiers

Chemical Name or Material Trifluoroacetaldehyde hydrate
CAS 421-53-4
Purity Grade Technical Grade
Molecular Formula C2H3F3O2
Linear Formula CF3CH(OH)2
Formula Weight 116.04 (Anhydrous)
MDL Number MFCD00167359
UN Number UN1989
Synonym 2,2,2-Trifluoro-1,1-ethanediol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1323409

  SDS
alfa aesar™
A1323409*
10g Each for $123.41
Description & Specifications

Specifications

Formula Weight 116.04 (Anhydrous)
Purity Grade Technical Grade
Assay 75%
Linear Formula CF3CH(OH)2
MDL Number MFCD00167359
CAS 421-53-4
UN Number UN1989
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H318-H302-H317-H335-H315
Highly flammable liquid and vapour.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number KI2850000
Concentration ≈75% in water
Boiling Point 104° to 106°C
Quantity 10g
Chemical Name or Material Trifluoroacetaldehyde hydrate
TSCA TSCA
Molecular Formula C2H3F3O2
Synonym 2,2,2-Trifluoro-1,1-ethanediol
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.

Applications
One general application of trifluoroacetaldehyde is the preparation of trifluoroethylamino derivatives via reductive amination reaction. This synthesis includes the formation of the corresponding N,O-acetal intermediates followed by their reduction using NaBH4 or 2-picoline borane complex, affording the target trifluoroethylamino compounds in moderate to good yields. Another general application of trifluoroacetaldehyde is the synthesis of chiral α-substituted trifluoroethylamino compounds.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Keep in cool place.

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H318-H302-H317-H335-H315
Highly flammable liquid and vapour.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.