Please login to your online account to display your discounted pricing

(α,α,α-Trifluoro-p-tolyl)acetic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC214370050

 View more versions of this product

Catalog No. AC214370050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 204.15
Physical Form Crystals or Crystalline Powder
Quantity 5g
Color White to very pale yellow
Melting Point 81° to 85°C
CAS Max % 100.0
Packaging Glass bottle
CAS 32857-62-8
Chemical Name or Material (α, α, α-Trifluoro-p-tolyl)acetic acid
Assay Percent Range 98%
Molecular Formula C9H7F3O2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
MDL Number MFCD00004352
CAS Min % 97.5
Linear Formula CF3C6H4CH2CO2H
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (α, α, α-Trifluoro-p-tolyl)acetic acid
CAS 32857-62-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H7F3O2
Linear Formula CF3C6H4CH2CO2H
Formula Weight 204.15
MDL Number MFCD00004352
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents