missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)phenol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 204780010

 View more versions of this product

Catalog No. AC204780010



Specifications

Specifications

2-(Trifluoromethyl)phenol
Authentic
White
1g
98.5
99%
CF3C6H4OH
06, III, 1263
ZOQOPXVJANRGJZ-UHFFFAOYSA-N
2-(trifluoromethyl)phenol
67958
Crystalline Low Melting Solid
65°C
Glass bottle
45°C to 46°C
444-30-4
100.0
C7H5F3O
MFCD00002222
2-hydroxy-alpha,alpha,alpha-trifluorotoluene, 2-hydroxybenzotrifluoride, 2-trifluoromethyl phenol, a,a,a-trifluoro-o-cresol, alpha,alpha,alpha-trifluoro-o-cresol, o-hydroxybenzotrifluoride, o-trifluoromethoxy phenol, o-trifluoromethylphenol, phenol, 2-trifluoromethyl
C1=CC=C(C(=C1)C(F)(F)F)O
162.11
162.11
99%

Chemical Identifiers

444-30-4
162.11
ZOQOPXVJANRGJZ-UHFFFAOYSA-N
67958
C1=CC=C(C(=C1)C(F)(F)F)O
C7H5F3O
MFCD00002222
2-hydroxy-alpha,alpha,alpha-trifluorotoluene, 2-hydroxybenzotrifluoride, 2-trifluoromethyl phenol, a,a,a-trifluoro-o-cresol, alpha,alpha,alpha-trifluoro-o-cresol, o-hydroxybenzotrifluoride, o-trifluoromethoxy phenol, o-trifluoromethylphenol, phenol, 2-trifluoromethyl
2-(trifluoromethyl)phenol
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 207-148-7

SDS Documents