Please login to your online account to display your discounted pricing

α,α,α-Trifluoro-m-tolyl isothiocyanate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material α, α, α-Trifluoro-m-tolyl isothiocyanate
CAS 1840-19-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H4F3NS
Linear Formula CF3C6H4NCS
Formula Weight 203.19
MDL Number MFCD00004806
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC306890050

  SDS
thermo fisher scientific
306890050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 203.19
Physical Form Liquid
Quantity 5g
Color Clear yellow to orange
Density 1.3350g/mL
CAS Max % 100.0
Molecular Formula C8H4F3NS
Assay Percent Range 98%
Chemical Name or Material α, α, α-Trifluoro-m-tolyl isothiocyanate
CAS 1840-19-3
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00004806
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
CAS Min % 97.5
Packaging Glass bottle
Synonym 3-Trifluoromethylphenylisothiocyanate
Refractive Index 1.5550 to 1.5570 (20°C, 589nm)
Linear Formula CF3C6H4NCS
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.