missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α,α,α-Trifluoro-m-tolunitrile, 96%, ACROS Organics™

$31.25 - $31.25

Chemical Identifiers

CAS 368-77-4
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C8H4F3N
Linear Formula CF3C6H4CN
Formula Weight 171.12
MDL Number MFCD00001807
Synonym 3-(Trifluoromethyl)benzonitrile
Chemical Name or Material α, α, α-Trifluoro-m-tolunitrile, 96%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC174190050 SDS Thermo Fisher Scientific
174190050
Glass bottle 5mL Each for $31.25
Specifications Description & Specifications

Chemical Identifiers

368-77-4
100.0
C8H4F3N
171.12
3-(Trifluoromethyl)benzonitrile
≥95%
95.0
96%
CF3C6H4CN
MFCD00001807
α, α, α-Trifluoro-m-tolunitrile, 96%

Specifications

1.2800g/mL
5mL
Authentic
100.0
1.4570 to 1.4610 (20°C, 589nm)
96%
CF3C6H4CN
MFCD00001807
3-(Trifluoromethyl)benzonitrile
≥95%
Colorless to yellow-orange
368-77-4
95.0
Glass bottle
Low Melting Solid
C8H4F3N
171.12
09, 478
α, α, α-Trifluoro-m-tolunitrile, 96%

Safety and Handling

GHS P Statement
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-711-4

TSCA : TSCA