missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α,α,α-Trifluoro-m-cresol, 98+%, ACROS Organics™

Manufacturer:  Acros Organics 168471000

 View more versions of this product

Catalog No. AC168471000


 

Specifications

Specifications

α, α, α-Trifluoro-m-cresol
Yellow
98+%
98.0
Authentic
1.4570 to 1.4590 (20°C, 589nm)
98+%
CF3C6H4OH
06, I, 187
UGEJOEBBMPOJMT-UHFFFAOYSA-N
3-(trifluoromethyl)phenol
7376
Liquid
1.3300g/mL
100mL
98-17-9
100.0
Glass bottle
≥98.0%
C7H5F3O
MFCD00002299
m-Hydroxybenzotrifluoride, m-Trifluoromethylphenol
C1=CC(=CC(=C1)O)C(F)(F)F
162.111
162.11

Chemical Identifiers

98-17-9
162.111
UGEJOEBBMPOJMT-UHFFFAOYSA-N
7376
C7H5F3O
MFCD00002299
m-Hydroxybenzotrifluoride, m-Trifluoromethylphenol
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 202-645-5

RTECSNumber : GP3510000

TSCA : TSCA

SDS Documents