Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethyltin chloride

$90.05 - $90.05

Chemical Identifiers

Chemical Name or Material Triethyltin chloride
CAS 994-31-0
Formula Weight 241.33
MDL Number MFCD00040414
UN Number UN2788
Synonym Chlorotriethylstannane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3960106

  SDS
alfa aesar™
3960106
5g Each for $90.05

AA3960103

  SDS
alfa aesar™
3960103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3960114

  SDS
alfa aesar™
3960114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 241.33
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Triethyltin chloride
Boiling Point 206°C
TSCA TSCA
Physical Form Liquid
MDL Number MFCD00040414
CAS 994-31-0
Quantity 5g
UN Number UN2788
Synonym Chlorotriethylstannane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Odor Unpleasant
Density 1.429g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Sensitivity Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.