missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethyltin bromide

Manufacturer:  Alfa Aesar™ 7113506

 View more versions of this product

Catalog No. AA7113506



Specifications

Specifications

Triethyltin bromide
-13.5°C
5g
1.63g/mL
Moisture Sensitive
MFCD00000052
triethyltin bromide, stannane, bromotriethyl, triethyl tin bromide, triethylstannium bromide, tin, triethyl-, bromide, unii-uop17166kc, triethylstannyl bromide, bromo triethyl stannane, stannane,bromotriethyl, acmc-1cl7y
CC[Sn](CC)(CC)Br
285.8
285.79
223°C to 225°C
2767-54-6
99°C (210°F)
1.520
C6H15BrSn
UN2788
KQPIFPBKXYBDGV-UHFFFAOYSA-M
bromo(triethyl)stannane
17701
Liquid

Chemical Identifiers

2767-54-6
285.8
KQPIFPBKXYBDGV-UHFFFAOYSA-M
17701
CC[Sn](CC)(CC)Br
C6H15BrSn
MFCD00000052
triethyltin bromide, stannane, bromotriethyl, triethyl tin bromide, triethylstannium bromide, tin, triethyl-, bromide, unii-uop17166kc, triethylstannyl bromide, bromo triethyl stannane, stannane,bromotriethyl, acmc-1cl7y
bromo(triethyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

TSCA : TSCA

SDS Documents