Please login to your online account to display your discounted pricing

(Triethylsilyl)acetylene 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (Triethylsilyl)acetylene
Assay Percent Range 97%
Molecular Formula C8H16Si
Formula Weight 140.3
MDL Number MFCD00075373
Synonym Ethynyltriethylsilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC368730050

  SDS
thermo fisher scientific
AC368730050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 140.3
Quantity 5g
Density 0.783g/cm3
Assay Percent Range 97%
Molecular Formula C8H16Si
Chemical Name or Material (Triethylsilyl)acetylene
MDL Number MFCD00075373
CAS 1777-03-3
Synonym Ethynyltriethylsilane