missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethylsilane, 99%, AcroSeal™, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC446161000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Triethylsilane | |
107°C to 108°C | |
98.5% min. (GC) | |
(C2H5)3SiH | |
15,9841 | |
Solubility in water: decomposes | |
CC[Si](CC)CC | |
116.28 | |
116.28 | |
AcroSeal™ Glass Bottle | |
0.72 |
617-86-7 | |
100 mL | |
C6H16Si | |
-3°C | |
c6h16si, hydride, silane e3h, silane, triethyl, silane, triethyl-,, triethylhydrosilane, triethylsilane, triethylsilane tes, triethylsilyl, triethylsilyl radical | |
QXTIBZLKQPJVII-UHFFFAOYSA-N | |
triethylsilicon | |
6327258 | |
99% | |
1.4100 to 1.412 | |
0.7200g/mL |
Chemical Identifiers
617-86-7 | |
116.28 | |
c6h16si, hydride, silane e3h, silane, triethyl, silane, triethyl-,, triethylhydrosilane, triethylsilane, triethylsilane tes, triethylsilyl, triethylsilyl radical | |
triethylsilicon |
C6H16Si | |
QXTIBZLKQPJVII-UHFFFAOYSA-N | |
6327258 | |
CC[Si](CC)CC |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 210-535-3
RUO – Research Use Only