missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylphosphine oxide, 98+%, ACROS Organics™

$93.79 - $93.79

Chemical Identifiers

CAS 597-50-2
Assay Percent Range 98+%
Molecular Formula C6H15OP
Formula Weight 134.16
MDL Number MFCD00013914
Chemical Name or Material Triethylphosphine oxide
Assay ≥98.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC380030050 SDS Thermo Fisher Scientific
AC380030050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC380030010 SDS Thermo Fisher Scientific
ACAC380030010
Glass bottle 1g Each for $93.79
Specifications Description & Specifications

Chemical Identifiers

597-50-2
C6H15OP
MFCD00013914
≥98.0%
98+%
134.16
Triethylphosphine oxide

Specifications

White
597-50-2
Glass bottle
98+%
134.16
Triethylphosphine oxide
5g
Authentic
Needles
C6H15OP
MFCD00013914
≥98.0%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger