missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethylphosphine oxide

Manufacturer:  Alfa Aesar™ 3039103

 View more versions of this product

Catalog No. AA3039103



Specifications

Specifications

Triethylphosphine oxide
48°C to 50°C
597-50-2
MFCD00013914
CCP(=O)(CC)CC
134.159
134.16
White
1g
C6H15OP
ZSSWXNPRLJLCDU-UHFFFAOYSA-N
1-diethylphosphorylethane
79061
Crystalline

Chemical Identifiers

597-50-2
134.159
ZSSWXNPRLJLCDU-UHFFFAOYSA-N
1-diethylphosphorylethane
C6H15OP
MFCD00013914
79061
CCP(=O)(CC)CC
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SZ1640000

SDS Documents