missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylphosphine, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar 3017714

 View more versions of this product

Catalog No. AA3017714



Specifications

Specifications

Triethylphosphine
Air & Moisture Sensitive
25g
97%
MFCD00009256
14,9675
RXJKFRMDXUJTEX-UHFFFAOYSA-N
triethylphosphane
27365
118.16
97%
0.81g/mL
126°C to 128°C
554-70-1
C6H15P
UN2845
triethylphosphine, phosphine, triethyl, triethylphosphorus, triethyl phosphine, pet3, et3p, unii-5w435d16pm, 3ep, triethylphophine, phosphorus triethyl
CCP(CC)CC
118.16
CHEBI:39971
Liquid

Chemical Identifiers

554-70-1
118.16
RXJKFRMDXUJTEX-UHFFFAOYSA-N
27365
triethylphosphane
C6H15P
MFCD00009256
triethylphosphine, phosphine, triethyl, triethylphosphorus, triethyl phosphine, pet3, et3p, unii-5w435d16pm, 3ep, triethylphophine, phosphorus triethyl
CHEBI:39971
CCP(CC)CC
Safety and Handling

Safety and Handling

GHS H Statement
H225-H250-H252-H332
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Harmful if inhaled.

GHS P Statement
P210-P222-P261-P303+P361+P353-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents