missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethyloxonium tetrafluoroborate, 95%, ACROS Organics™

Chemical Identifiers

CAS 368-39-8
Molecular Formula C6H15O·BF4
Molecular Weight (g/mol) 189.99
InChI Key IYDQMLLDOVRSJJ-UHFFFAOYSA-N
Synonym boron tetrafluoride triethyl oxonium, meerwein's reagent, triethoxonium fluoroborate, triethyloxidanium tetrafluoroborate, triethyloxonium borofluoride, triethyloxonium fluoborate, triethyloxonium fluoroborate, triethyloxonium tetraflouroborate, triethyloxonium tetrafluoroborate, unii-z0b19dd36j
PubChem CID 2723982
IUPAC Name triethyloxidanium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CC[O+](CC)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC436760250
SDS View Documents
Thermo Fisher Scientific
AC436760250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC436761000
SDS View Documents
Thermo Fisher Scientific
AC436761000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Alkyl transfer reagent
Specifications

Chemical Identifiers

368-39-8
189.99
boron tetrafluoride triethyl oxonium, meerwein's reagent, triethoxonium fluoroborate, triethyloxidanium tetrafluoroborate, triethyloxonium borofluoride, triethyloxonium fluoborate, triethyloxonium fluoroborate, triethyloxonium tetraflouroborate, triethyloxonium tetrafluoroborate, unii-z0b19dd36j
triethyloxidanium;tetrafluoroborate
C6H15O·BF4
IYDQMLLDOVRSJJ-UHFFFAOYSA-N
2723982
[B-](F)(F)(F)F.CC[O+](CC)CC

Specifications

Glass bottle
White
25g
95%
15,5867
Solubility in water: decomposes
[B-](F)(F)(F)F.CC[O+](CC)CC
189.99
189.99
95%
3 to 5 % ether
84.0°C to 89.0°C
368-39-8,60-29-7
C6H15O·BF4
boron tetrafluoride triethyl oxonium, meerwein's reagent, triethoxonium fluoroborate, triethyloxidanium tetrafluoroborate, triethyloxonium borofluoride, triethyloxonium fluoborate, triethyloxonium fluoroborate, triethyloxonium tetraflouroborate, triethyloxonium tetrafluoroborate, unii-z0b19dd36j
IYDQMLLDOVRSJJ-UHFFFAOYSA-N
triethyloxidanium;tetrafluoroborate
2723982
Powder
Triethyloxonium tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-705-1

SDS