missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride, ACROS Organics™

Manufacturer:  Acros Organics 175111000

 View more versions of this product

Catalog No. AC175111000


 

Description

Description

Alkylation reagent
Specifications

Specifications

Triethyloxonium tetrafluoroborate
368-39-8,75-09-2
(C2H5)3OBF4
1.3280g/mL
IYDQMLLDOVRSJJ-UHFFFAOYSA-N
triethyloxidanium;tetrafluoroborate
2723982
Plastic bottle
1.328
1M Solution in Methylene Chloride
C6H15BF4O
MFCD00044423
01,1210; 02,430; 03,303; 04,527; 05,691; 06,611; 07,386; 08,500; 09,482; 10,417; 15,337
[B-](F)(F)(F)F.CC[O+](CC)CC
189.989
189.99
Solubility in water: reacts.
100mL

Chemical Identifiers

368-39-8
189.989
IYDQMLLDOVRSJJ-UHFFFAOYSA-N
C6H15BF4O
MFCD00044423
2723982
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing cancer.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 206-705-1

TSCA : TSCA

SDS Documents