Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethyloxonium tetrafluoroborate, 1.0M in dichloromethane, packaged under Argon in resealable ChemSeal™ bottles

Chemical Identifiers

Chemical Name or Material Triethyloxonium tetrafluoroborate
Linear Formula (CH3CH2)3OBF4
Formula Weight 189.99
MDL Number MFCD00044423
UN Number UN2922
Beilstein 3598090
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42934AE

 
alfa aesar™
42934AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 189.99
Chemical Name or Material Triethyloxonium tetrafluoroborate
TSCA TSCA
Density 1.328g/mL
MDL Number MFCD00044423
Beilstein 3598090
Health Hazard 1 Danger
Packaging ChemSeal™ bottles
Health Hazard 2 GHS H Statement
H314-H318-H351
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 −30° to -10°C
Color Colorless
DOT Information Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
RTECS Number RR4584700
Concentration 1.0M in dichloromethane
Quantity 100mL
Physical Form Liquid
Linear Formula (CH3CH2)3OBF4
UN Number UN2922
Odor Ether-like
Sensitivity Moisture Sensitive

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H351
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.

−30° to -10°C