missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylmethylammonium Chloride 98.0+%, TCI America™

Manufacturer:  TCI America M0852500G

 View more versions of this product

Catalog No. M0852500G



Specifications

Specifications

Triethylmethylammonium Chloride
500g
C7H18ClN
acmc-1bu0o
CC[N+](C)(CC)CC.[Cl-]
151.678
151.68
≥98.0% (T)
White
10052-47-8
MFCD00059972
NIUZJTWSUGSWJI-UHFFFAOYSA-M
triethyl(methyl)azanium;chloride
82326
Crystal-Powder at 20°C

Chemical Identifiers

10052-47-8
151.678
NIUZJTWSUGSWJI-UHFFFAOYSA-M
82326
CC[N+](C)(CC)CC.[Cl-]
C7H18ClN
MFCD00059972
acmc-1bu0o
triethyl(methyl)azanium;chloride
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

TSCA : Yes

Documents