Please login to your online account to display your discounted pricing

Triethylenetetramine (Technical), Fisher Chemical


Quantity 500mL
Packaging Glass Bottle

Manufacturer: fisher scientific company  BPT410-500

 View more versions of this product

Catalog No. T410-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Density 0.980g/cm³
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure <0.01 hPa at 20�C
Boiling Point 280°C / 536°F
pH 14
Melting Point -35°C / -31°F
Color Light Yellow
Quantity 500mL
Chemical Name or Material Triethylenetetramine
CAS 112-24-3
Purity Grade Technical
Physical Form Liquid
Molecular Formula C6H18N4
Formula Weight 146.24g/mol
MDL Number MFCD00008169
Synonym TETA
ChemAlert Storage Symbol White
DOT Information DOT Class 8, : Corrosive

Specifications

Chemical Name or Material Triethylenetetramine
CAS 112-24-3
Purity Grade Technical
Molecular Formula C6H18N4
Formula Weight 146.24g/mol
MDL Number MFCD00008169
Synonym TETA
Accessories
 

SDS & Safety Documents