Please login to your online account to display your discounted pricing

Triethylenetetramine (Technical), Fisher Chemical


Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  T4101

 View more versions of this product

Catalog No. T410-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 146.24
Boiling Point 280°C
Melting Point −35°C
Vapor Pressure <0.01 hPa at 20°C
Quantity 1L
DOT Information DOT Class 8, : Corrosive
pH 14
Density 0.980g/cm³
Physical Form Liquid
Molecular Formula C6H18N4
Color Light Yellow
Chemical Name or Material Triethylenetetramine
Identification Pass Test
MDL Number MFCD00008169
ChemAlert Storage Symbol White
Synonym TETA
Packaging Glass Bottle
CAS 112-24-3

Specifications

Chemical Name or Material Triethylenetetramine
CAS 112-24-3
Purity Grade Technical Grade
Molecular Formula C6H18N4
Formula Weight 146.24
MDL Number MFCD00008169
Synonym TETA
Accessories
 

SDS & Safety Documents