Please login to your online account to display your discounted pricing

Triethylenetetramine (Technical), Fisher Chemical

$279.93 - $279.93

Chemical Identifiers

Chemical Name or Material Triethylenetetramine
CAS 112-24-3
Purity Grade Technical Grade
Molecular Formula C6H18N4
Formula Weight 146.24
MDL Number MFCD00008169
Synonym TETA
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

T410-1

  SDS
fisher scientific company
T4101
Glass Bottle 1L Each for $279.93
Case of 6 Each for $1,679.58
Description & Specifications

Specifications

Chemical Name or Material Triethylenetetramine
Quantity 1L
CAS 112-24-3
ChemAlert Storage Symbol White
DOT Information DOT Class 8, : Corrosive
Density 0.980g/cm³
Purity Grade Technical Grade
Physical Form Liquid
Identification Pass Test
Molecular Formula C6H18N4
Formula Weight 146.24
MDL Number MFCD00008169
Synonym TETA
Packaging Glass Bottle
Vapor Pressure <0.01 hPa at 20°C
Boiling Point 280°C
pH 14
Melting Point −35°C
Color Light Yellow

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling