missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethylene glycol monobutyl ether, 98%

$47.26 - $110.39

Chemical Identifiers

CAS 143-22-6
Molecular Formula CH3(CH2)3O(CH2CH2O)3H
Formula Weight 206.28
MDL Number MFCD00020606
Beilstein 1750600
Chemical Name or Material Triethylene glycol monobutyl ether
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL047380B SDS Alfa Aesar™
L047380B
1000g Each for $110.39
AAL0473830 SDS Alfa Aesar™
L0473830
250g Each for $47.26
Specifications

Description

Applications
Triethylene glycol monobutyl ether, glycol ether a highly effective carrier solvent for textile dye processes. With superior surface tension characteristics, water solubility and solvency for oils, it has potential for use in household, institutional, industrial and special-purpose cleaners. The coupling ability of this product enhances performance and improves shelf stability of cleaning products. It is also used as an active solvent for solvent-based coatings, dye carrier for textile dye processes, coupling agent and solvent in household and industrial cleaners, paint and floor polish strippers, hard surface cleaners, and disinfectants, chemical process solvent and intermediate for ester production used as solvents, surfactants and plasticizers, coupling agent for resins and dyes in water-based printing ink, component of high-boiling hydraulic brake fluids.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Description & Specifications

Chemical Identifiers

143-22-6
206.28
1750600
CH3(CH2)3O(CH2CH2O)3H
MFCD00020606
Triethylene glycol monobutyl ether

Specifications

1000g
CH3(CH2)3O(CH2CH2O)3H
MFCD00020606
272°C
147°C (297°F)
98%
143-22-6
206.28
1750600
−35°C
Triethylene glycol monobutyl ether

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

RTECSNumber : KJ9450000

TSCA : TSCA