missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylene Glycol Monobenzyl Ether, TCI America™

Manufacturer:  TCI America T199625G

 View more versions of this product

Catalog No. T199625G



Specifications

Specifications

Triethylene Glycol Monobenzyl Ether
25g
C13H20O4
2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethanol
C1=CC=C(C=C1)COCCOCCOCCO
240.299
240.30
≥97.0% (GC)
Yellow
55489-58-2
MFCD02093556
KVPHZILZNXDCNH-UHFFFAOYSA-N
2-[2-(2-phenylmethoxyethoxy)ethoxy]ethanol
2737322
Clear Liquid at 20°C

Chemical Identifiers

55489-58-2
240.299
KVPHZILZNXDCNH-UHFFFAOYSA-N
2737322
C1=CC=C(C=C1)COCCOCCOCCO
C13H20O4
MFCD02093556
2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethanol
2-[2-(2-phenylmethoxyethoxy)ethoxy]ethanol
Safety and Handling

Safety and Handling

TSCA : No

Documents