missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylene Glycol Mono(2-propynyl) Ether, TCI America™

Manufacturer:  TCI America T31145G

 View more versions of this product

Catalog No. T31145G



Specifications

Specifications

Triethylene Glycol Mono(2-propynyl) Ether
5g
C9H16O4
2-[2-[2-(2-Propynyloxy)ethoxy]ethoxy]ethanol, Triethylene Glycol Monopropargyl Ether
C#CCOCCOCCOCCO
188.223
188.22
≥96.0% (GC)
Yellow
208827-90-1
MFCD13189984
CXJWUJYYDLYCCQ-UHFFFAOYSA-N
2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethanol
11506589
Clear Liquid at 20°C

Chemical Identifiers

208827-90-1
188.223
CXJWUJYYDLYCCQ-UHFFFAOYSA-N
11506589
C#CCOCCOCCOCCO
C9H16O4
MFCD13189984
2-[2-[2-(2-Propynyloxy)ethoxy]ethoxy]ethanol, Triethylene Glycol Monopropargyl Ether
2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethanol
Safety and Handling

Safety and Handling

TSCA : No

Documents